Glas-Col® Stirmantle Dual Control, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Combination stirring and temperature controller for Glas-Col stirmantles is often used to simultaneously power the Stirmantle's heating and stirring, as well as, an additional top mantle for those applications where vapor condensation on the upper half of the flask is undesirable.
- Two-outlet version allows up to 1800 W
- Three-outlet version allows up to 2400 W
Dual control features a universal J, T, and K probe input, a thermocouple break alarm, NEMA 5 - 15R receptacles, and a lab frame support bracket.
Customers Who Bought This Also Bought