Aven ProVue Solas Magnifying Lamp, Aven
Catalog # 76289-866
Supplier: Aven Tools
Specifications
- Magnification:2.25×
- Description:ProVue Solas Magnifying Lamp
- Cat. no.:76289-866
Specifications
About this item
Offers flexibility of using the 5-Diopter Lens (2.25× magnification).
- 60 ultra bright SMD LEDs with brightness controls
- Fully enclosed spring balanced arm
- Protective lens cover
- Heavy duty table clamp
Optional 3-Diopter Lens (1.75× magnification, sold separately).
Customers Who Bought This Also Bought