Protocult Stool Collection Device, Therapak
Catalogue # 76203-712
Fournisseur: Therapak, LLC
Spécifications
- Description:Protocult stool collection device
- Dimensions:305×170 mm
- Cat. no.:76203-712
- Weight:17.5 g
Spécifications
À propos de cet article
This paper folding collection device consists of a disposable, water-resistant frame which supports an attached tear-away paper collection bowl as well as removable adhesive strips on either side allowing device to be securely anchored to commode.
- Pre-printed with easy to follow instructions
- Lightweight and ships flat