Cole-Parmer® XRF Prep-Aid® Grinding Aids and Sample Binders
Fournisseur: SPEX
Certificats
A propos de cet article
Prepare samples for reproducible and contamination-free results.
- Select binders from powder, tablet, or liquid form depending on your application
High-quality sample preparation is the key to achieving consistent and reliable analytical results. Powders are ideal for samples that crumble or ablate after pressing. They blend quickly and completely with samples to form XRF (x-ray fluorescence) sample disks. Tablets are an efficient way to add binder to XRF sample disks. Liquid easily mixes with the sample.
Customers Who Bought This Also Bought