ContactUsLinkComponent
Marina® Brush Kit
Marina® Brush Kit
# de catalogue 470308-612
Brush Kit Marina Aquarium With 3 Types
Marina® Brush Kit
# de catalogue 470308-612
Spécifications
  • Description:
    3 Brush Kit
  • Cat. no.:
    470308-612
Spécifications
A propos de cet article
The Marina Aquarium Brush Cleaning Kit contains three cleaning brushes.
The foxtail brush is ideal for cleaning tubes up to 2,54 cm (1") in diameter and 45 cm (18") in length. The airline brush is designed for cleaning airline tubing. The filter stem brush is great for cleaning external filter tubes as well as clip-on filter intake stems.
Customers Who Bought This Also Bought