Specifications
- Volume:1 oz.
- Description:Biological Enhancer
- Cat. no.:470308-828
Specifications
About this item
Fluval Biological Enhancer is a responsive biological aquarium supplement that immediately inoculates aquarium water with a powerful team of beneficial bacteria, providing a safe biological habitat for your fish.
- For fresh and salt water aquariums
- Powerful team of beneficial bacteria immediately inoculates aquarium water
- All-natural product
- Every container contains pure, safe bacterial colonies, free of undesirable contaminants
- Does not require refrigeration
It goes to work fast, releasing massive amounts of beneficial bacteria that eliminate toxic ammonia and nitrites while creating a biologically well-balanced aquarium for healthy fish to thrive.
Fluval Biological Enhancer is strongly recommended for new aquariums, as they do not contain the necessary concentrations of beneficial bacteria required to metabolize the waste that fish produce. It quickly establishes safe and essential conditions in new aquarium set-ups, so that you can introduce fish to new aquariums immediately.
Regular application helps to competitively exclude the establishment of undesirable bacteria in aquariums. Fluval Biological Enhancer provides consistent results thanks to our unique and proprietary bio-floc technology that ensures product stability, efficacy and shelf life. High-quality production processes ensure that every container contains pure, safe bacterial colonies, free of undesirable contaminants.