Adhesive Paper Labels 'Exempt Specimen', Labelmaster
# de catalogue 101377-351
Fournisseur: Label Master
Spécifications
- Color:Red
- Description:Adhesive paper labels
- Material:Semi-Gloss Paper
- Thickness:5.85 Mil
- Cat. no.:101377-351
- Legend:Exempt Human Specimen
- Length:1.375"
- Width:2.25"
- Supplier No.:L358
Spécifications
A propos de cet article
These paper labels can be used for shipping medical specimens.
Comply with the requirements in International Air Transport, ICAO and DOT for marking the package orientation. The bright red label surface has chemical and abrasion resistance, and the backing offers strong adhesion to paint, steel, fiber and plastic surfaces.
Customers Who Bought This Also Bought