Exo-Terra® Coco Husk Substrate
# de catalogue 470308-876
Fournisseur: ROLF C. HAGEN (USA) CORP.
Spécifications
- Description:Cocohusk Brick
- Cat. no.:470308-876
Spécifications
A propos de cet article
Easy to use substrate.
- 100% natural substrate
- Ideal for natural terrarium set-ups
- Increases humidity in terrariums
- Stimulates natural digging and burrowing behavior
Exo Terra Coco Husk is made from compressed coconut husk from the plantations of tropical Asia. This ecological substrate is ideal for increasing humidity in the terrarium and is totally safe for frogs, salamanders and burrowing or digging animals.
Customers Who Bought This Also Bought