Cole-Parmer® Deuterium Lamps
Supplier: Cole-Parmer
Certificates
About this item
Economical replacement deuterium lamps.
- Select from standard or long life lamps
- Manufactured in an ISO accredited facility
- Individually tested and issued with a Test Certificate
This range of deuterium (D2) lamps are made to fit the majority of detectors used in analytical labs today. The lamps are used by many of the major pharmaceutical and contract research organizations (CROs) around the globe.
These deuterium lamps are manufactured under stringent ISO 9001 quality procedures. Each lamp is manufactured to rigorous standards and traceable from manufacturing to customer delivery.
Customers Who Bought This Also Bought