Hydrion® Lo Iodine Test Kit, Micro Essential Laboratory
Catalogue # 89370-912
Fournisseur: Micro Essential
Numéro CAS:
Spécifications
- Description:Hydrion Lo Iodine Test Kit
- Cat. no.:89370-912
- Supplier No.:IL-250
Spécifications
À propos de cet article
The Hydrion Lo iodine kit provides an easy way to estimate the amount of titratable iodine in sanitizing solutions made with commercial iodophors. The color chart is easy to read and has color matches at 0, 12.5, 25, 50 ppm (parts per million) of titratable iodine.
Delivery information: Each kit contains one 15 foot roll of iodine test paper, matching color chart, and two sample tubes with caps. In addition there is one extra roll of iodine test paper and a laminated color chart. Provides for approximately 200 tests.