A propos de cet article
Everything you'll need to recreate a successful marine reef.
- Low energy LED lighting
- Includes hardware needed for setup
Designed with clean lines and a contemporary finish, the Fluval Reef aquaria and stand sets come equipped with everything you'll need to recreate a successful marine reef at home, including Fluval's widely popular Marine and Reef Performance LED. These high-efficiency lights feature 6 unique LED band widths with essential actinic blue wavelengths for full spectrum coverage that will drive photosynthetic activity and coral growth, while also offering an accurate viewing experience. The glass aquarium features a unique frosted rim frame that keeps unsightly water lines out of view, while the elegant two-tone cabinet is constructed from high-quality laminated wood. The set also comes equipped with an effective CP Circulation Pump. All other hardware is neatly concealable within the cabinet.
Stylish reef aquarium and cabinet set Glass aquarium with frosted rim framing and a high-quality laminated cabinet with two-tone doors Integrated compartment houses a filter and Fluval Sea Protein Skimmer for outstanding filtration Practical drain valve with drain hose make water changes so easy and convenient Sleek and adjustable Fluval Sea Marine & Reef Performance LED Lighting promotes and supports strong coral growth and color Powerful day & night illumination with 3 settings: ON (day, night) and OFF switch All hardware is neatly concealed in the cabinet so that you can enjoy a pleasing aesthetic experience.