Magid® ComfortFlex® Polyethylene Powder-Free Disposable Glove, Magid
Catalog # 69100-094
Supplier: Magid Glove
Small Business Enterprise
Specifications
- Glove Color:Translucent
- Glove size:L
- Ambidextrous/Hand-specific:Ambidextrous
- Disposable / reusable:Disposable
- Latex/Latex-free:Latex-free
- Powdered/Powder-free:Powder-free
- Description:Magid® ComfortFlex® Polyethylene Powder-Free Disposable Glove
- Length:26.5 cm (10.5")
- Finish:Textured
- Cat. no.:69100-094
- Supplier No.:H99BOXEMBL
Specifications
About this item
Gloves are fast and easy to slip on and off and are the most economical choice.
- Textured high-density polyethylene with strong seams for durability
- One size fits all
For single-use applications where gloves are worn for a short amount of time, especially for food handling.
Certifications: Complies with FDA food handling requirements (21 CFR 177.2600).
Packaging: 100 gloves per dispenser.
Customers Who Bought This Also Bought