ContactUsLinkComponent
Hood with Faceshield, Magid Glove
Hood with Faceshield, Magid Glove
# de catalogue 69100-136
Fournisseur:  Magid Glove
Small Business Enterprise
Hood with Faceshield, Magid Glove
# de catalogue 69100-136
Fournisseur:  Magid Glove
Small Business Enterprise
Spécifications
  • Garment application:
    Cleanroom
  • Sterile:
    +
  • Color:
    White
  • Cat. no.:
    69100-136
  • Supplier No.:
    CH14
Spécifications
A propos de cet article
Disposable hood that completely covers head and neck.
  • Hood is shoulder length and has two air holes around the chin
  • Opening around eye area for goggle use under hood that is surrounded with elastic seam
Customers Who Bought This Also Bought