Magid® EconoWear® Knee-High Tyvek® Boot Covers, Magid Glove
# de catalogue 32893-196
Fournisseur: Magid Glove
Small Business Enterprise
Spécifications
- Garment application:Cleanroom
- Sterile:+
- Cat. no.:32893-196
- Supplier No.:SC116-XL
Spécifications
A propos de cet article
Shoe covers offer the protection of Tyvek in a disposable, low cost solution.
- Knee-high shoe/boot covers have yellow, vinyl non-skid soles
- Skid-resistant
Tyvek fabric is very strong, is nearly impossible for the average person to tear, but can easily be cut with scissors or other sharp objects.
Packaging: 25 per pack.
Customers Who Bought This Also Bought