Magid® EconoWear® Tyvek® Bib Apron, Magid Glove
# de catalogue 32893-184
Fournisseur: Magid Glove
Small Business Enterprise
Spécifications
- Garment application:Industrial
- W×L:28×36"
- Sterile:+
- Color:White
- Fabric:Tyvek®
- Cat. no.:32893-184
- Supplier No.:A32
Spécifications
A propos de cet article
Aprons provide protection against tears, punctures, liquid spills and splashes.
- Bib-style apron
- Binding ties around waist
Tyvek fabric is very strong, it is nearly impossible for the average person to tear, but can easily be cut with scissors or other sharp objects.
Packaging: 100/ea per case.
Customers Who Bought This Also Bought