ContactUsLinkComponent
Magid® EconoWear Lite N Kool Disposable Jacket, Magid Glove
Magid® EconoWear Lite N Kool Disposable Jacket, Magid Glove
# de catalogue 32893-144
Fournisseur:  Magid Glove
Small Business Enterprise
Magid® EconoWear Lite N Kool Disposable Jacket, Magid Glove
# de catalogue 32893-144
Fournisseur:  Magid Glove
Small Business Enterprise
Spécifications
  • Clothing Size:
    XL
  • Garment application:
    Industrial
  • Color:
    White
  • Cat. no.:
    32893-144
  • Supplier No.:
    J91-XL
Spécifications
A propos de cet article
EconoWear Lite N Kool jacket provides protection against punctures, liquid spills and splashes.
  • Manufactured from tear and puncture resistant spun bonded polypropylene
  • Jackets have a snap front
The jackets made for general applications where economy, coolness, and comfort are top priority.
Certifications: Manufactured to ANSI/ISEA spec. 101-1993 standards.
Customers Who Bought This Also Bought