Magid® EconoWear Lite N Kool Disposable Jacket, Magid Glove
# de catalogue 32893-144
Fournisseur: Magid Glove
Small Business Enterprise
Spécifications
- Clothing Size:XL
- Garment application:Industrial
- Color:White
- Cat. no.:32893-144
- Supplier No.:J91-XL
Spécifications
A propos de cet article
EconoWear Lite N Kool jacket provides protection against punctures, liquid spills and splashes.
- Manufactured from tear and puncture resistant spun bonded polypropylene
- Jackets have a snap front
The jackets made for general applications where economy, coolness, and comfort are top priority.
Certifications: Manufactured to ANSI/ISEA spec. 101-1993 standards.
Customers Who Bought This Also Bought