ContactUsLinkComponent
Round Steel 7 Hole Lockout, Magid Glove
Round Steel 7 Hole Lockout, Magid Glove
# de catalogue 69100-518
Fournisseur:  Magid Glove
Small Business Enterprise
Round Steel 7 Hole Lockout, Magid Glove
# de catalogue 69100-518
Fournisseur:  Magid Glove
Small Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Color:
    Silver
  • Description:
    7 Hole Lockout
  • Cat. no.:
    69100-518
  • Supplier No.:
    DM15072
Spécifications
A propos de cet article
Made of heavy stamped steel, lockout hasp allows for lockout of the same piece of equipment by up to 7 workers.
The 11/2" jaw accommodates padlocks up to 63/8". Tamper-resistant scissor-style means security and durability.
Customers Who Bought This Also Bought