DuPont™ Tyvek® Two-Pocket Jacket, Magid Glove
Fournisseur: Magid Glove
Certificats
A propos de cet article
The EconoWear jacket provides protection against punctures, liquid spills and splashes.
- Jackets have a snap front
Tyvek fabric is very strong, but is nearly impossible for the average person to tear, but can easily be cut with scissors or other sharp objects.
Certifications: Manufactured to ANSI/ISEA specifications 101-1993 standards.
Packaging: 50 per case.
Spécifications
- Catalog No:
- 32893-140
- 32893-142
- Fabric:
- Tyvek
- Tyvek
- Seam type:
- Serged
- Serged
- Garment application:
- Industrial
- Industrial