Magid® Wool Lined Heat Gloves, Magid
Fournisseur: Magid Glove
Certificats
A propos de cet article
100% Kevlar® wool lined gloves are abrasion, cut and heat resistant.
- Extra wide cuff opening for easy on and off
- High tensile strength protects hands during hazardous applications up to 800 °F
- Lightweight canvas weave that provides maximum flexibility
Use in glass and sheet metal manufacturing/handling, steel mills and foundries.
Spécifications
- Catalog No:
- 11000-281
- Ambidextrous/Hand-specific:
- Hand-specific
- Disposable / reusable:
- Reusable
- Latex/Latex-free:
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Material:
- Kevlar®