Ward's® Professional Insect Net
# de catalogue 470308-231
Fournisseur: Shiv Dial Sud & Sons
New Product
Spécifications
- Description:Professional insect net set
Spécifications
A propos de cet article
A favorite of collectors for over 50 years.
- Component assembly
- Crafted for multiple uses
- Strong and durable
- Nets are 550 micron (µm) mesh
This professional insect net package is just what you need as a teacher. This package is assembled for basic aerial sweep collection. The heavy-duty 29.5" metal handle is longer than the student version. The metal handle has a larger diameter for strength, a high-gloss finish, and marine brass connection system. The 12" steel ring connects to the metal handle using pins and a retaining ring. The net easily slides onto the ring using a muslin sleeve and can be easily changed for a different style of net.
BodyTextAccessories: Replacement rings and nets can also be seen on our 'Build Your Own Insect Net' page.
Customers Who Bought This Also Bought