ContactUsLinkComponent
NF-κB (human p50) Transcription Factor Assay Kit, Cayman Chemical Company
NF-κB (human p50) Transcription Factor Assay Kit, Cayman Chemical Company
# de catalogue 75816-692
Numéro CAS:  
NF-KB (human p50) Transcription Factor Assay Kit
NF-κB (human p50) Transcription Factor Assay Kit, Cayman Chemical Company
# de catalogue 75816-692
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    NF-κB (human p50) Transcription Factor Assay Kit
  • No. of wells:
    96
  • Cat. no.:
    75816-692
  • Contents:
    Kit contains: Assay Buffer, Reagents, Positive Control, Antibody Binding Buffer, Primary Antibody, Buffer Concentrate, Polysorbate 20, NF-kB Competitor dsDNA, Goat Anti-Rabbit HRP Conjugate, 96-well Strip Plate, 96-well Cover Sheet, Developing Solution, and Stop Solution
  • Supplier No.:
    10006912-96
Spécifications
A propos de cet article
A 96-well assay for measurement of NF-κB (human p50).
  • A sensitive, non-radioactive method of detecting NF-κB (p50) from whole cell lysates
  • 96-well plate format replaces EMSAs
  • Capture the transcription factor using a specific dsDNA sequence bound to the plate
  • Detect the dsDNA-bound transcription factor with specific antibodies in an ELISA format
Nuclear Extraction Kit available to aid in the isolation of nuclear and cytoplasmic fractions from cell lysates or tissue homogenates.
Cayman’s NF-κB (human p50) Transcription Factor Assay is a non-radioactive, sensitive method for detecting specific transcription factor DNA binding activity in nuclear extracts and whole cell lysates.
A specific double stranded DNA (dsDNA) sequence containing the NF-κB response element is immobilized to the wells of a 96-well plate. NF-κB contained in a nuclear extract, binds specifically to the NF-κB response element.
NF-κB (p50) is detected by addition of specific primary antibody directed against NF-κB (p50). A secondary antibody conjugated to HRP is added to provide a sensitive colorimetric readout at 450 nm.
The Cayman Chemical NF-κB (human p50) Transcription Factor Assay detects human NF-κB (p50). It will not cross-react with NF-κB (p65).
Customers Who Bought This Also Bought