Magid® CutMaster® Medium Weight Kevlar® Leather Palm Gloves, Magid
Fournisseur: Magid Glove
Certificats
A propos de cet article
This gloves combines the toughness and durability of a split leather palm with a 100% Kevlar machine knit shell.
- Quality cow split leather palm is sewn on using Kevlar thread to ensure a long-lasting seam
- Color-coded overedge wrist prevents unraveling and keeps a tight fit after washing
Kevlar fiber is five times stronger than steel and gives this glove superior ANSI cut Level 4 protection.
Spécifications
- Catalog No:
- 69100-600
- 69100-168
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free