Magid® KnitMaster White Standard Weight Machine Knit Gloves, Magid
# de catalogue 11000-282
Fournisseur: Magid Glove
Small Business Enterprise
Spécifications
- Ambidextrous/Hand-specific:Ambidextrous
- Glove Color:White
- Glove size:M
- Length:21.6 cm (8.5")
- Powdered/Powder-free:Powder-free
- Disposable / reusable:Reusable
- Latex/Latex-free:Latex-free
- Gauge:7
- Cat. no.:11000-282
- Supplier No.:T193C
Spécifications
A propos de cet article
An economical alternative to standard weight canvas and jersey gloves, the Magid KnitMaster insulated work gloves are seamless, reversible cotton/polyester knit gloves.
- White cotton polyester knit glove
- Reversible and long lasting
- Great insulating liner
- Color-coded wrist edge
These long-lasting material handling gloves feature a white threaded wrist edge to prevent unraveling when the glove is washed. The versatile gloves is a perfect glove for general maintenance, packaging, inspection, material handling, assembly, and shipping and receiving. These grip gloves also serve as a great insulating liner for warmth.
Customers Who Bought This Also Bought