Magid® DuraMaster® Gunn Cut Cow Split Leather Palm, Magid
# de catalogue 32889-332
Fournisseur: Magid Glove
Small Business Enterprise
Spécifications
- Ambidextrous/Hand-specific:Hand-specific
- Glove Color:Gray
- Glove size:L
- Powdered/Powder-free:Powder-free
- Description:Magid® DuraMaster® Gunn Cut Cow Split Leather Palm
- Disposable / reusable:Reusable
- Latex/Latex-free:Latex-free
- Cat. no.:32889-332
- Supplier No.:AWT20
Spécifications
A propos de cet article
Gunn cut pattern work gloves employ a shoulder split leather palm and full leather index fingers and tips to provide protection where needed most.
- Shoulder split leather palm and full leather index fingers and tips
- Palm lining
Gloves are pearl colored with striped back and white cuff.
Customers Who Bought This Also Bought