Magid® CleanMaster® Men's and Women's Nylon Gloves, Magid
Fournisseur: Magid Glove
Certificats
A propos de cet article
Magid's stretch nylon gloves are soft and durable, perfect for inspection or light work.
- Two-piece stretch nylon gloves
- Low linting for less contamination
- Color-coded hem for easy matching
These gloves feature a reversible cut and sewn style, allowing them to fit either hand. This feature ensures a longer life for the inspection gloves, as well as improved ease in sorting them. These gloves also feature a hemmed edge to provide the ultimate fit and are available in multiple lengths for additional coverage.
Spécifications
- Catalog No:
- 32889-320
- 32889-336
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Material:
- Nylon
- Nylon