Magid® CutMaster® Kevlar® Dotted Knit Gloves, Magid
Supplier: Magid Glove
Certificates
About this item
Suited especially for glass and metal handling or assembly, provides excellent all-purpose hand protection with added grip and dexterity provided by the glove's double sided plastic dot coating.
- 100% Kevlar® composition
- Black plastic dots on both sides
- Seamless, reversible machine knit design
Pound for pound, Kevlar fiber has been shown to be five times stronger than steel, making it an ideal material in the manufacture of cut resistance work gloves. These gloves features a color coded overcast rubber band prevents unraveling and allows for easy size identification for dispensing and for sorting after laundering.
Specifications
- Catalog No:
- 69100-590
- 69100-592
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Gauge:
- 7
- 7