Magid® TouchMaster® 650H/651H Lightweight Cotton Lisle Inspection Glove, Magid
Supplier: Magid Glove
Certificates
About this item
The inspection glove is 100% cotton two-piece lisle glove constructed of cool and breathable cotton.
- Two-piece cotton construction
- Cut and sewn pattern
- Color-coded sizes for easy sorting
- Hemmed edge to prevent unraveling
This glove is form fitting and it features a hemmed edge to prevent unraveling in the wash. The glove can be worn alone to protect delicate products from smudges, finger prints or skin oils, or it can be used as a liner to absorb moisture. This glove is a great general-purpose glove for light jobs in any industry.
Specifications
- Catalog No:
- 11000-252
- 11000-296
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Material:
- Cotton
- Cotton