Magid® MultiMaster® Double Palm Glove with Band Top Cuff, Magid
# de catalogue 69100-020
Fournisseur: Magid Glove
Small Business Enterprise
Spécifications
- Ambidextrous/Hand-specific:Hand-specific
- Glove Color:Yellow
- Glove size:L
- Powdered/Powder-free:Powder-free
- Description:Magid® MultiMaster® Double Palm Glove with Band Top Cuff
- Disposable / reusable:Reusable
- Latex/Latex-free:Latex-free
- Material:Cotton/polyester
- Cuff length:21/2"
- Cat. no.:69100-020
- Packaging:12/Dozen
- Supplier No.:64BT
Spécifications
A propos de cet article
Magid® MultiMaster® fleece glove feature a clute pattern with two layers of 100% cotton fabric on the palm, making them perfect durable work gloves for jobs requiring good thermal protection and durability.
- 100% cotton fabric
- Easy-fitting clute-cut pattern
- Band top cuff for easy on/off
The nap-out fabric gives these textile gloves fantastic heat protection and grip, making a great economical choice for general purpose hand protection.
Customers Who Bought This Also Bought