Magid® TouchMaster® Lightweight Knit Lisle Gloves, Magid
Fournisseur: Magid Glove
Certificats
A propos de cet article
Magid® TouchMaster® machine-knit lisle gloves fits like a dress glove. These lightweight, seamless style has a reversible pattern for longer wear. This glove is knit from a cotton and polyester blend providing cool and comfortable fit.
- Knit wrist cuff for a secure fit
- Color-coded sizes for easy sorting
- Breathable lightweight fabric
These long-lasting gloves are also feature with an over-edge hem to ensure a snug fit and prevent unraveling when washed. These gloves are perfect for inspection or any general-purpose use.
Spécifications
- Catalog No:
- 61300-100
- 11000-258
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Material:
- Cotton/polyester
- Cotton/polyester
- Gauge:
- 13
- 13
- Sterility:
- Non sterile
- Non sterile