Magid® MultiMaster® Double Palm Glove with Band Top Cuff, Magid
Fournisseur: Magid Glove
Certificats
A propos de cet article
Magid® MultiMaster® Double palm gloves are features with a clute pattern with two layers of 100% cotton fabric in vital wear areas such as the palm, thumb and the back of the index finger.
- Easy-fitting clute-cut pattern
- 18 oz.
This glove provides a perfect balance of cost-savings and performance, also featuring a 2" band top cuff for easy on and off. The nap-in fabric gives this glove added durability, making this as a great, economical choice for general purpose hand protection.
Spécifications
- Catalog No:
- 39100-124
- 39100-122
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Material:
- Cotton/polyester
- Cotton/polyester