Magid® ROC® Polyurethane Palm Coated Gloves, Magid
Fournisseur: Magid Glove
Certificats
A propos de cet article
Magid® ROC® gloves are flexible and form fitting with polyurethane coated palms. These are feature with a seamless machine knit, 100% nylon shell with a knit wrist cuff.
- Breathable
- Scratch-resistant polyurethane palm coating
- Comfortable knit wrist cuff
- Designed for lint free applications
The polyurethane palm coating provides excellent grip, abrasion resistance and dexterity for precise and delicate operations. These useful white gloves are perfect for electronics, cleanroom and paint departments where lint is problematic.
Spécifications
- Catalog No:
- 46620-050
- 46620-049
- 46620-047
- 46620-045
- 46620-043
- Model:
- PU50
- PU50
- PU50
- PU50
- PU50
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nylon/PU
- Nylon/PU
- Nylon/PU
- Nylon/PU
- Nylon/PU
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile