Magid® ROC® Polyurethane Palm Coated Gloves, Magid
Fournisseur: Magid Glove
Certificats
A propos de cet article
Work Gloves feature a durable, silicone free, polyurethane palm coating that provides excellent grip in either oily or dry conditions.
- Breathable, form-fitting, 100% stretch nylon shell
These gloves with gauge, seamless, machine knit shell and knit wrist cuff, are form-fitting delivering superb comfort and breathability during even the longest shift.
The ROC® Glove's nylon shell is also tough providing good abrasion resistance and ANSI Cut Level 1 protection. Ideal choice in hand protection for general maintenance, assembly, and material handling.With the ROC, it feels like working with your bare hands.
Packaging: Packed one dozen to a polybag.
Spécifications
- Catalog No:
- 69100-434
- 69100-432
- 69100-430
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nylon
- Nylon
- Nylon
- Gauge:
- 15
- 15
- 15
- Cuff style:
- Knit Wrist
- Knit Wrist
- Knit Wrist