DuPont™ Tyvek® Smock with Waist Ties, Magid Glove
Fournisseur: Magid Glove
Certificats
A propos de cet article
Smocks provide protection against tears, punctures, liquid spills and splashes.
- Self ties around the waist
- Long sleeves
Tyvek fabric is very strong, that is impossible to tear by average person, but can easily be cut with scissors or other sharp objects. These smocks are 41" in length.
Packaging: 50 per case.
Spécifications
- Catalog No:
- 69100-076
- 69100-074
- 69100-080
- 69100-078
- Garment application:
- Medical, Clinical, Laboratory
- Medical, Clinical, Laboratory
- Medical, Clinical, Laboratory
- Medical, Clinical, Laboratory