Magid® Thermal Leather Firefighters Gloves, Magid
Fournisseur: Magid Glove
Certificats
A propos de cet article
Magid® leather firefighter gloves are manufactured from specially treated and tanned thermal leather which makes the glove to be highly heat resistant and added a feature such as good abrasion and cut resistance.
- High heat and water-resistant
- Gunn cut pattern
- Kevlar knit wrist cuff
- Wing thumb type
These gloves feature with a 2" turtleneck for additional protection. Due to their high heat resistance, these gloves are generally recommended for applications of 500 to 800 °F. The closed wool lining provides more heat protection than fleece lining.
Spécifications
- Catalog No:
- 11000-238
- 11000-240
- 11000-242
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- Wool
- Wool
- Wool
- Material:
- Leather, premium split cowhide
- Leather, premium split cowhide
- Leather, premium split cowhide
- Cuff style:
- Knit Wrist
- Knit Wrist
- Knit Wrist