Magid® TouchMaster® Heavyweight Lisle Gloves, Magid
Fournisseur: Magid Glove
Certificats
A propos de cet article
Magid's TouchMaster is a two-piece lisle inspection glove constructed of cool and breathable cotton.
- Cut and sewn pattern
- Color-coded sizes for easy sorting
- Breathable lightweight fabric
- 100% cotton
- Optional hemmed edge to prevent unraveling
Faetures form fitting, heavyweight for the most comfortable fit. This glove can be worn alone to protect delicate products from smudges, finger prints or skin oils, or it can be used as a liner to absorb moisture. General-purpose glove for light jobs in any industry.
Spécifications
- Catalog No:
- 32889-298
- 32889-299
- 11000-250
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Material:
- Cotton
- Cotton
- Cotton