Magid® TouchMaster® Inspection Gloves, Magid
Fournisseur: Magid Glove
Certificats
A propos de cet article
Medium weight cotton lisle inspection gloves are 100% cotton two-piece lisle inspection glove constructed of cool and breathable bleached white cotton.
- Form-fitting and cut and sewn for the most comfortable fit
- Color-coded sizes for easy sorting
They can be worn alone to protect delicate products from smudges, finger prints or skin oils, or used as a liner to absorb moisture. They are an excellent choice for jobs in the plastics industry.
Spécifications
- Catalog No:
- 11000-265
- 32889-460
- 11000-294
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free