Magid® CleanMaster® Nylon Tricot Gloves, Magid
Fournisseur: Magid Glove
Certificats
A propos de cet article
These gloves are comfortable and durable for cleanroom, paint and electronics projects.
- Two-piece nylon tricot glove
- Low linting for less contamination
- Color-coded hem for easy matching
This style is hemmed, with a two-piece cut and sewn pattern. These gloves are reversible and fits either hand, ensuring longer wear and greater ease in sorting after laundry.
An insert thumb provides a better fit, giving this comfortable style the wear of a dress glove.
Spécifications
- Catalog No:
- 32889-308
- 32889-310
- 32889-312
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nylon
- Nylon
- Nylon