Electronic Overhead Stirrers, VELP Scientifica
Fournisseur: VELP SCIENTIFIC INC.
Certificats
A propos de cet article
Complete range of overhead stirrers with electronic adjustment for a variety of needs in terms of viscosity and volume.
- Wide range of applications: solubilization, mixing, emulsification and homogenization
- Self-locking chuck simplifies assembly
- Gentle start-up ensures optimum progression of the stirring speed
- Most advanced safety standards
- Through-hole to accept long shafts
Overhead stirrers with a technopolymer structure, ideal for premium resistance to acids, bases and solvents. Many reliable solutions are available, according to different requirements in terms of viscosity and volume. All the models are equipped with a user-friendly self-locking chuck, that simplifies assembly and the gentle start-up ensures optimum progression of the stirring speed. Ensures the most advanced safety standards.
Certifications: Meets CE, UL, and CAN/CSA electrical safety requirements.
BodyTextAccessories: Accessories not included, unless stated in description. Accessories can be purchased separately.
Spécifications
- Catalog No:
- 76288-118
- 76288-120
- 76288-116
- 76288-172
- 76288-174
- 76288-176
- 76288-178
- Speed range:
- 50 - 2000 rpm, with SpeedServo™ to ensure constant speed and timer
- 50 - 2000 rpm
- max 1300 rpm
- 20 - 1200 rpm
- 50 - 2000 rpm
- 50 - 2000 rpm
- 50 - 2000 rpm, with SpeedServo™ to ensure constant speed and timer