Magid® Greyt Shadow® Guard Standard Weight Terrycloth Glove, Magid
# de catalogue 69100-017
Fournisseur: Magid Glove
Small Business Enterprise
Spécifications
- Ambidextrous/Hand-specific:Ambidextrous
- Glove Color:Gray
- Glove size:L
- Length:25.4 cm (10")
- Powdered/Powder-free:Powder-free
- Description:Magid® Greyt Shadow® Guard Standard Weight Terrycloth Glove
- Disposable / reusable:Reusable
- Latex/Latex-free:Latex-free
- Material:Cotton/polyester
- Sterility:Non sterile
- Gauge:7
- Cat. no.:69100-017
- Supplier No.:GT948R
Spécifications
A propos de cet article
The Magid Greyt shadow guard terrycloth machine knit work gloves offer good thermal protection and strong cut and abrasion resistance with form-fitting comfort.
- Gray 60% cotton and 40% polyester medium heavyweight blend
- Seamless, reversible, machine knit construction fits either hand
- Good cut and abrasion resistance
- Gray color extends life of the glove
The glove's gray color effectively hides dirt and grime reducing the rate of disposal and extending the life of these heat protection gloves. The Greyt shadow guard cotton inspection gloves fit either hand, simplifying sorting and dispensing.
The glove's extra tight, high density, knit construction offers improved comfort, dexterity and longer wear.
Customers Who Bought This Also Bought