Vaplock™ Safety Waste Funnel, Cole-Parmer
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Vaplock™ SF Series funnels allow for safer disposal of pourable laboratory waste.
Vaplock™ SF Series funnels allow for safer disposal of pourable laboratory waste.
As with all solvent applications, chemical compatibility should be noted prior to purchase.
A locking, hinged lid minimizes leaks and spills when the funnel is closed. If used in tandem with other Vaplock™ products, such as a manifold and exhaust filter, a closed system may be maintained for passive wastes while the funnel is sealed.
Customers Who Bought This Also Bought