Vaplock™ DIN 50 Waste Caps and Waste Kits, Cole-Parmer
Fournisseur: Argos Technologies
Certificats
A propos de cet article
The Vaplock™ closed system design reduces chemical evaporation and exposure to hazardous vapors.
- Minimize exposure to harmful chemical vapors
- Collect solvent waste from multiple effluent streams
- Highly adaptable and conformable to a broad range of containment types
Featuring a vapor tight connection, waste manifolds are designed to fit standard DIN 50 threaded lab bottles. Chemical-resistant materials of construction include polypropylene or PTFE manifold and insert, with polypropylene spin collar. These versatile manifolds can be configured for barbed connections, compression fittings, 1/4-28 fittings, and ferrule sets. All manifolds are designed to accommodate the EF-100 Filter and indicator.
Ordering information: Kits include (1) 10-port waste manifold, (3) 1/4" NPT polypropylene plugs, (6) 1/4-28 PTFE plugs, (1) 1/2" NPT polypropylene plug, (1) organic vapor filter, (2) 2.2 mm O.D. hard wall tubing connections, (1) 2.5 mm O.D. hard wall tubing connection, (3) 1/8" O.D. hard wall tubing connections, (2) 1/16" O.D. hard wall tubing connections, (3) barbed tubing connection 1/4" I.D. tubing×1/4" NPT polypropylene, (1) barbed tubing connection 1/8" I.D. tubing×1/4" NPT polypropylene, (1) barbed tubing connection 3/16" I.D. tubing×1/4" NPT polypropylene, (1) barbed tubing connection 3/8" I.D. tubing×1/4" NPT polypropylene, (1) barb to barb Y connector with 1/4" (6.25 mm) barbs, polypropylene, (1) hard wall tube extension with union connections and 5' of 1/8" O.D. PTFE tubing, (1) filter adapter 1/4" NPT×1/2" NPT polypropylene, and (1) breakthrough color indicator for Vaplock™ organic vapor filter.
Container not included.
Container not included.
Customers Who Bought This Also Bought