Airfree® Schlenk Vacuum Manifolds, Double Bank, Inert Gas, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Manifolds are modular in design, allowing the user tremendous flexibility in performing experiments.
- Forward facing 0 to 8 mm Chem-Vac™ Chem-Cap®
- Main ports are spaced 100 mm center to center, with an additional 2 ports on the upper bank
- All connection ports are #15 O-Ring joint connections that incorporate a greaseless O-Ring seal
Double bank manifold available in 3, 4, or 5 port configurations. Forward facing 0 to 8 mm Chem-Vac™ Chem-Cap® valves permit easy operation inside of a fume hood. Main ports are spaced 100 mm center to center, with an additional 2 ports on the upper bank that can be used for gauging or sealed with stoppers.
Adapters from #15 O-Ring joints to standard taper components, serrated hose connections, or 3/8" Cajon® fittings are available to allow the ports to be used with existing glassware. See (17003-150) for replacement Chem-Vac™, Chem-Cap® plugs.
Customers Who Bought This Also Bought