Adjustable Height Probe Holders, for Bioreactors, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Probe holders for 12 mm or 19 mm probes.
- Adjustable height with a compression fitting
Serve as replacement parts for Applikon® Biotechnology and Broadley-James® brands of bioreactors.
Customers Who Bought This Also Bought