J-KEM® Temperature Controllers, Model 150, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
An ideal controller for applications that do not require the precise regulation of the J-KEM® 200 series controllers.
- Advanced PID algorithm
- 1200 watts of power
This compact unit packs sufficient for 5 l heating mantles, many ovens, and other devices. Built with the J-KEM® state-of-the-art PID process controller, these units do not contain the J-KEM® power control computer.
Each instrument is individually calibrated against National Institute of Standards and Technology Traceable equipment.
Certifications: NIST traceable.
Customers Who Bought This Also Bought