Ward's® Silicone Lab Mat
# de catalogue 470306-958
Fournisseur: Heathrow Scientific
Spécifications
- Dimensions:350×600×20 mm
- Color:Blue/white
- Cat. no.:470306-958
Spécifications
A propos de cet article
Mat features magnetic area to keep stir bars in place. It keeps benchtop clean and safe from stains, spills and wear.
- Reusable
- Easy to clean and store
Anti-skid working surface keeps items in place and reduces sound and vibration from equipment.
Customers Who Bought This Also Bought