A propos de cet article
Lance MT can be used for the simulation of finger stick and blood glucose testing. Practice without the pain.
This product will enable a student or patient to have a realistic experience when testing blood sugar, using a glucose meter, and obtaining a reading. Lance fits over the finger of a standardized patient, or patient simulator, and has a protective liner that will prevent the actual finger from being pierced with the lancing device. "Lance MT" is shipped empty and can be filled with a control solution matching the glucose meter being used. This option allows users to use their meter, and get a reading with the control solution being the Blood in the simulated finger.
Blood sample, and proceed in the simulation with either a simulated blood glucose machine (not included), or an instructor provided simulated blood glucose level.