Vacuum Check Valve, Chemglass
# de catalogue 80072-684
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Description:Vacuum check valve
- Cat. no.:80072-684
- Length:125 mm
- Supplier No.:CG-4539-01
Spécifications
A propos de cet article
Valve has an ¹/₄" O.Ø precision ground ceramic ball which fits into the ground seat to effect a liquid seal.
- Tubing O.Ø: 10 mm
Customers Who Bought This Also Bought