ContactUsLinkComponent
DNA/RNA Shield™ Blood Collection Tube, Zymo Research
DNA/RNA Shield™ Blood Collection Tube, Zymo Research
Catalog # 76020-434
Supplier:  Zymo Research
DNA/RNA Shield* - Blood Collection Tube 16 mm x 100 mm
DNA/RNA Shield™ Blood Collection Tube, Zymo Research
Catalog # 76020-434
Supplier:  Zymo Research
Specifications
  • Ø:
    16 mm
  • Volume:
    3 ml (blood draw)
  • Description:
    DNA/RNA Shield™ blood collection tube
  • Length:
    100 mm
  • Cat. no.:
    76020-434
  • Tube Dimensions:
    16×100 mm (prefilled with DNA/RNA shield, 6 ml)
  • Additive:
    Prefilled with DNA/RNA shield, 6 ml
Specifications
About this item
Conveniently collect whole blood directly into DNA/RNA Shield™ blood vacuum tubes. Each evacuated tube draws in an exact amount of whole blood which instantly stabilizes the nucleic acid and inactivates any harmful/pathogenic organisms in the sample.
  • Nucleic acid preservation (at ambient temperaturea and cold-free)
  • Pathogen inactivation (bacteria, fungus, parasites and viruses)
  • Streamlined purification (no reagent removal, universally compatible, automatable)
DNA/RNA Shield™ blood vacuum tubes applicable for next-generation sequencing, qPCR and microarray.
Customers Who Bought This Also Bought