A propos de cet article
Gravity convection laboratory ovens are recommended for critical scientific and analytical use in laboratory and pilot plant product research. Typical applications include aging, curing, drying, baking, pre-heating, conditioning and life cycle testing.
- Fast ramp-up to 306 °C for faster cycle times
- Temperature range approximately 20 °C above ambient to 306 °C for multiple applications
- Advanced PID temperature control system for sensitive response
- Built-in digital timer for automatic operation following manual start
- Independent over temperature set-point and operational control override for additional safety
- Interior construction with 430 series stainless steel
- Stainless steel shelves are adjustable on 0.5" (12.7 mm) centers to provide flexible inventory options
SGO Series ovens are available in 110 to 120 V, 220 to 240 V, 50/60 Hz configurations. These feature forced-air circulation to assure maximum temperature uniformity, triple-wall construction to minimize external surface temperature.
Rear access port, 1.75" diameter (44 mm) for independent cables, instrumentation, Exhaust port, 1.75" diameter (44 mm) adjustable, with external connection to customer-supplied vent for effluent exhaust if required.
Upper vents are closed when recovery times following door openings are tested. Published recovery performance is based on a standard cabinet under controlled testing at a (22 ± 3 °C ambient temperature, line voltage ±10% of specified voltage.
Spécifications
- Catalog No:
- 76210-100
- 76210-384
- Temperature range:
- 20 °C above ambient to 306 °C
- 20 °C above ambient to 306 °C
- Temperature uniformity:
- 2.5 (80 °C);3.5 (150 °C);4.0 (306 °C)
- 2.5 (80 °C);3.5 (150 °C);4.0 (306 °C)