Corepoint Scientific™ General Purpose Undercounter Refrigerator and Freezer, Horizon Scientific
Fournisseur: Horizon Scientific
Certificats
A propos de cet article
These undercounter refrigerators and freezers provide laboratory cold storage for general purpose applications.
- Keyed door locks
- Mechanical thermostats
- Interior shelving
- Magnetic door gaskets
- Natural hydrocarbon refrigerants
These units provide multi-functional features in cost-effective designs. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Ordering information: All units have a one-year parts and labor warranty.
Spécifications
- Catalog No:
- 76271-350
- 76271-348
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76271-350
- 76271-348
- Defrost:
- Manual
- Cycle
- Shelves/drawers:
- 2 fixed shelves with 3 removable drawers
- 2 adjustable shelves, 1 fixed shelf
- Door type:
- 1 solid swing door, hinge right
- 1 solid swing door, hinge right
- Electrical:
- 115 V, 60 Hz, 0.9 A, ¹/₇ HP
- 115 V, 60 Hz, 0.9 A, ¹/₇ HP
- Environmental certification:
- HFC Free
- HFC Free